missing translation for 'onlineSavingsMsg'
Learn More

3-[(4-Chlorobenzyl)thio]-1H-1,2,4-triazole, Alfa Aesar™

Product Code. 11430170
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430170 250 mg 250mg
11440170 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430170 Supplier Thermo Scientific Alfa Aesar Supplier No. H50647.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 134796-34-2
Molecular Formula C9H8ClN3S
Molecular Weight (g/mol) 225.69
MDL Number MFCD01211119
InChI Key IPYLPWUQKVUWGY-UHFFFAOYSA-N
Synonym 3-4-chlorobenzyl thio-1h-1,2,4-triazole, 3-4-chlorophenyl methyl sulfanyl-1h-1,2,4-triazole, 3-4-chloro-benzylsulfanyl-4h-1,2,4 triazole, 3-4-chlorobenzyl sulfanyl-4h-1,2,4-triazole, 5-4-chlorophenyl methylsulfanyl-1h-1,2,4-triazole, 3-4-chlorophenyl methylthio-1h-1,2,4-triazole, 3-4-chlorophenyl methyl sulfanyl-2h-1,2,4-triazole, cbmicro_005601, cambridge id 5854799
PubChem CID 719011
IUPAC Name 5-[(4-chlorophenyl)methylsulfanyl]-1H-1,2,4-triazole
SMILES ClC1=CC=C(CSC2=NC=NN2)C=C1

Specifications

Quantity 250 mg
Formula Weight 225.7
Chemical Name or Material 3-[(4-Chlorobenzyl)thio]-1H-1,2,4-triazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.