missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Chlorobenzal)phthalide, TRC

Product Code. 30133230
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30133230 10 g 10g
30007225 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133230 Supplier Toronto Research Chemicals Supplier No. TRCC36451510G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 20526-97-0
Molecular Formula C15 H9 Cl O2
Molecular Weight (g/mol) 256.68
Synonym 1(3H)-Isobenzofuranone, 3-[(4-chlorophenyl)methylene]-, Phthalide, 3-(p-chlorobenzylidene)- (6CI,7CI,8CI), 3-(4-Chlorobenzal)phthalide, 3-(4-Chlorobenzylidene)-2-benzofuran-1(3H)-one, 3-(p-Chlorobenzylidene)phthalide, Azelastine Related Compound E (USP)
IUPAC Name 3-[(4-chlorophenyl)methylidene]-2-benzofuran-1-one
SMILES Clc1ccc(C=C2OC(=O)c3ccccc23)cc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C15H9ClO2/c16-11-7-5-10(6-8-11)9-14-12-3-1-2-4-13(12)15(17)18-14/h1-9H
Formula Weight 256.0291
Percent Purity >95%
Chemical Name or Material 3-(4-Chlorobenzylidene)isobenzofuran-1(3H)-one
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.