missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Bromophenylthio)propionic acid, 98%, Thermo Scientific™

Product Code. 15444207
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
This item is not returnable. View return policy

Product Code. 15444207

Brand: Thermo Scientific Alfa Aesar H31609.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13735-04-1
Molecular Formula C9H9BrO2S
Molecular Weight (g/mol) 261.133
MDL Number MFCD05664871
InChI Key CGKAMPCZJIBUGT-UHFFFAOYSA-N
Synonym 3-4-bromophenyl sulfanyl propanoic acid, 3-4-bromophenyl thio propanoic acid, 3-4-bromophenylthio propanoic acid, 3-4-bromophenylthio propionic acid, 3-4-bromothiophenoxy propionic acid
PubChem CID 16776242
IUPAC Name 3-(4-bromophenyl)sulfanylpropanoic acid
SMILES C1=CC(=CC=C1SCCC(=O)O)Br

Specifications

Melting Point 109°C to 112°C
Quantity 10 g
Formula Weight 261.14
Percent Purity 98%
Chemical Name or Material 3-(4-Bromophenylthio)propionic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.