missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Bromophenoxy)-6-methylpyridazine, 97%, Thermo Scientific™

Product Code. p-2699074
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10202352

Brand: Thermo Scientific Maybridge CC21310CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 368869-96-9
Molecular Formula C11H9BrN2O
Molecular Weight (g/mol) 265.11
MDL Number MFCD03086135
InChI Key LZEJOINDIKGWQJ-UHFFFAOYSA-N
Synonym 3-4-bromophenoxy-6-methylpyridazine, pyridazine,3-4-bromophenoxy-6-methyl, 3-4-bromanylphenoxy-6-methyl-pyridazine, 4-bromo-1-6-methylpyridazin-3-yloxy benzene
PubChem CID 2776522
IUPAC Name 3-(4-bromophenoxy)-6-methylpyridazine
SMILES CC1=NN=C(OC2=CC=C(Br)C=C2)C=C1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 265.11
Percent Purity 95%
Chemical Name or Material 3-(4-Bromophenoxy)-6-methylpyridazine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.