missing translation for 'onlineSavingsMsg'
Learn More

3-[4-(Bromomethyl)phenyl]-5-methyl-1,2,4-oxadiazole 97+%, Thermo Scientific™

Product Code. 10148042
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10148042 1 g Amber Glass Bottle 1g
10269382 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10148042 Supplier Thermo Scientific Maybridge Supplier No. CC34808DA
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 256956-42-0
Molecular Formula C10H9BrN2O
Molecular Weight (g/mol) 253.099
InChI Key BMXSEDLZMUPKSH-UHFFFAOYSA-N
Synonym 3-4-bromomethyl phenyl-5-methyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 3-4-bromomethyl phenyl-5-methyl, 1,2,4-oxadiazole,3-4-bromomethyl phenyl-5-methyl, 3-4-bromomethyl phenyl-5-methyl-1,2,4-oxadiazol
PubChem CID 7164590
IUPAC Name 3-[4-(bromomethyl)phenyl]-5-methyl-1,2,4-oxadiazole
SMILES CC1=NC(=NO1)C2=CC=C(C=C2)CBr

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 253.1
Percent Purity 97%
Chemical Name or Material 3-[4-(Bromomethyl)phenyl]-5-methyl-1, 2, 4-oxadiazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.