missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Biphenylyl)-D-alanine, 98%, Thermo Scientific™

Product Code. 15440669
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 170080-13-4
Molecular Formula C15H15NO2
Molecular Weight (g/mol) 241.29
MDL Number MFCD01318879
InChI Key JCZLABDVDPYLRZ-CQSZACIVSA-N
Synonym r-3-1,1'-biphenyl-4-yl-2-aminopropanoic acid, d-4,4'-biphenylalanine, h-4-phenyl-d-phe-oh, 4-phenyl-d-phenylalanine, d-biphenylalanine, 4-phenyl-d-phenyalanine, d-4,4'-biphenylphenylalanine, h-d-bip-oh, 4'-biphenyl-d-ala, 2r-2-amino-3-4-phenylphenyl propanoic acid
PubChem CID 7006711
IUPAC Name (2R)-2-amino-3-(4-phenylphenyl)propanoic acid
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)CC(C(=O)O)N

Specifications

Color White
Solubility Information Slightly soluble in water.
Formula Weight 241.29
Percent Purity 98%
Quantity 250 mg
Physical Form Powder
Chemical Name or Material 3-(4-Biphenylyl)-D-alanine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'