missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Aminophenyl)propionic Acid, TRC

Product Code. 30341549
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
Product Code. Quantity unitSize
30341549 2.5 g 2.5g
1 options
This item is not returnable. View return policy

Product Code. 30341549

Brand: Toronto Research Chemicals TRCA6379082.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(4-Aminophenyl)propionic Acid
CAS 2393-17-1
Molecular Formula C9H11NO2
Quantity 2.5 g
Synonym 3-(p-Aminophenyl)propanoic Acid, 3-(p-Aminophenyl)propionic Acid, 4-Aminohydrocinnamic Acid, NSC 25083, p-Aminohydrocinnamic Acid, beta-(4-Aminophenyl)propionic Acid
InChI Formula InChI=1S/C9H11NO2/c10-8-4-1-7(2-5-8)3-6-9(11)12/h1-2,4-5H,3,6,10H2,(H,11,12)
SMILES Nc1ccc(CCC(=O)O)cc1
IUPAC Name 3-(4-aminophenyl)propanoic acid
Molecular Weight (g/mol) 165.19
Formula Weight 165.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.