missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Aminobutyl)-1H-indole-5-carbonitrile, TRC

Product Code. 30403648
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30403648 50 mg
30393534 100 mg
30403073 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30403648 Supplier Toronto Research Chemicals Supplier No. TRCI62904850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143612-85-5
Molecular Formula C13H15N3
Molecular Weight (g/mol) 213.28
IUPAC Name 3-(4-aminobutyl)-1H-indole-5-carbonitrile
SMILES NCCCCc1c[nH]c2ccc(cc12)C#N

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H15N3/c14-6-2-1-3-11-9-16-13-5-4-10(8-15)7-12(11)13/h4-5,7,9,16H,1-3,6,14H2
Formula Weight 213.1266
Chemical Name or Material 3-(4-Aminobutyl)-1H-indole-5-carbonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.