Learn More
3,4,9,10-Perylenetetracarboxylic diimide, Thermo Scientific Chemicals
Description
Used in chemical research. PTCDI is used extensively as an industrial pigment. It is used as tunable laser dye, light-harvesting material, transistor, molecular switched, solar cell, and optoelectronic device. Planar perylene- and naphthalene-based diimide linkers can be employed to tether the Watson- Crick and the Hoogsteen strands of a DNA triplex, thus providing conjugates capable of targeting singlestranded nucleic acids with the formation of hairpin triplexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Quantity | 25 g |
Solubility Information | Insoluble in water. |
Formula Weight | 390.35 |
Physical Form | Powder |
Chemical Name or Material | 3,4,9,10-Perylenetetracarboxylic diimide |
RUO – Research Use Only