missing translation for 'onlineSavingsMsg'
Learn More

3,4,7,8-Tetramethyl-1,10-phenanthroline, 98+%

Product Code. 11341297
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11341297 5 g 5g
11331297 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11341297 Supplier Thermo Scientific Alfa Aesar Supplier No. L01482.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1660-93-1 | C16H16N2 | 236.318 g/mol

3,4,7,8-Tetramethyl-1,10-phenanthroline acts as a metal-chelating agent for transition metal-catalyzed processes. It is also used in the preparation of heteroleptic cationic iridium(III) complex viz. 3,4,7,8-tetramethyl-1,10-phenanthroline-bis[2-(2',4'-difluorophenyl)pyridine]iridium(III) hexafluorophosphate. Further, it is employed as a ligand and forms dinuclear copper(II) hypocrellin B complexes. In addition to this, it plays an important role in the formation of tetraaqua(3,4,7,8-tetramethyl-1,10-phenanthroline-kappa2N,N')zinc(II) thiosulfate complex.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,7,8-Tetramethyl-1,10-phenanthroline acts as a metal-chelating agent for transition metal-catalyzed processes. It is also used in the preparation of heteroleptic cationic iridium(III) complex viz. 3,4,7,8-tetramethyl-1,10-phenanthroline-bis[2-(2′,4′-difluorophenyl)pyridine]iridium(III) hexafluorophosphate. Further, it is employed as a ligand and forms dinuclear copper(II) hypocrellin B complexes. In addition to this, it plays an important role in the formation of tetraaqua(3,4,7,8-tetramethyl-1,10-phenanthroline-kappa2N,N′)zinc(II) thiosulfate complex.

Solubility
Soluble in 1,4-dioxane, acetone, dichloromethane, N,N-dimethylformamide, N,N-dimethylacetamide, methanol and hot toluene.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1660-93-1
Molecular Formula C16H16N2
Molecular Weight (g/mol) 236.318
MDL Number MFCD00004974
InChI Key NPAXPTHCUCUHPT-UHFFFAOYSA-N
Synonym 1,10-phenanthroline, 3,4,7,8-tetramethyl, 3,4,7,8-tetramethylphenanthroline, 3,4,7,8-tetramethyl-1, 10-phenanthroline, chembl99384, 3,4,7,8-tetramethyl-1,10 phenanthroline, 3,4,7,8-tetramethylpyridino 3,2-h quinoline, acmc-209dtn, 3,4,7,8-me4phenan, bidd:gt0406, 3,4,7,8-tetramethylphenantroline
PubChem CID 74265
IUPAC Name 3,4,7,8-tetramethyl-1,10-phenanthroline
SMILES CC1=CN=C2C(=C1C)C=CC3=C(C(=CN=C32)C)C

Specifications

Melting Point ∽280°C (decomposition)
Quantity 5 g
Beilstein 171279
Solubility Information Soluble in 1,4-dioxane,acetone,dichloromethane,N,N-dimethylformamide,N,N-dimethylacetamide,methanol and hot toluene.
Formula Weight 236.32
Percent Purity ≥98%
Chemical Name or Material 3,4,7,8-Tetramethyl-1,10-phenanthroline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.