missing translation for 'onlineSavingsMsg'
Learn More

3,4,6-Tri-O-tert-butyldimethylsilyl-D-glucal, 97%, Thermo Scientific™

Product Code. 15406758
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15406758 1 g 1g
15416758 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15406758 Supplier Thermo Scientific Alfa Aesar Supplier No. H60275.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents. Air, water, moisture sensitive..
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79999-47-6
Molecular Formula C24H52O4Si3
Molecular Weight (g/mol) 488.931
MDL Number MFCD00171721
InChI Key FMXIGEXRLSNRMR-NJYVYQBISA-N
Synonym tri-o-tert-butyldimethylsilyl-d-glucal, 3,4,6-tri-o-t-butyl-dimethylsilyl-d-glucal, 3-o,4-o,6-o-tris tert-butyldimethylsilyl-1,2-dideoxy-d-arabino-1-hexenopyranose, 2r,3r,4r-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-3,4-diyl bis oxy bis tert-butyldimethylsilane, tert-butyl 2r,3r,4r-4-tert-butyldimethylsilyl oxy-2-tert-butyldimethylsilyl oxy methyl-3,4-dihydro-2h-pyran-3-yl oxy dimethylsilane
PubChem CID 10648718
IUPAC Name [(2R,3R,4R)-3,4-bis[[tert-butyl(dimethyl)silyl]oxy]-3,4-dihydro-2H-pyran-2-yl]methoxy-tert-butyl-dimethylsilane
SMILES CC(C)(C)[Si](C)(C)OCC1C(C(C=CO1)O[Si](C)(C)C(C)(C)C)O[Si](C)(C)C(C)(C)C

Specifications

Density 0.928
Boiling Point 210°C (1.2 mmHg)
Flash Point >113°C (235°F)
Refractive Index 1.456
Quantity 1 g
Sensitivity Air and moisture sensitive
Optical Rotation −30° (c=1 in Chloroform)
Formula Weight 488.92
Percent Purity 97%
Chemical Name or Material 3,4,6-Tri-O-tert-butyldimethylsilyl-D-glucal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.