missing translation for 'onlineSavingsMsg'
Learn More

3,4,6-Tri-O-acetyl-D-galactal

Product Code. 15418459
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15418459 1 g 1g
15428459 5 g 5g
15438459 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15418459 Supplier Thermo Scientific Alfa Aesar Supplier No. J66904.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4098-06-0 | C12H16O7 | 272.25 g/mol

3,4,6-Tri-O-acetyl-D-galactal is important building block for both solution- and solid-phase synthesis of oligosaccharides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,6-Tri-O-acetyl-D-galactal is important building block for both solution- and solid-phase synthesis of oligosaccharides.

Solubility
Soluble in chloroform at 10mg/ml

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4098-06-0
Molecular Formula C12H16O7
Molecular Weight (g/mol) 272.25
MDL Number MFCD00064092
InChI Key LLPWGHLVUPBSLP-UHFFFAOYNA-N
Synonym 3,4,6-tri-o-acetyl-d-galactal, tri-o-acetyl-d-galactal, 2r,3r,4r-2-acetoxymethyl-3,4-dihydro-2h-pyran-3,4-diyl diacetate, d-arabino-hex-5-enitol, 2,6-anhydro-5-deoxy-, triacetate, 1,3,4-tri-o-acetyl-2,6-anhydro-5-deoxyhex-5-enitol, d-galactal triacetate, 2,6-anhydro-5-deoxy-d-arabino-hex-5-enitol triacetate, 2r,3r,4r-3,4-bis acetyloxy-3,4-dihydro-2h-pyran-2-yl methyl acetate, galactal triacetate, tri-o-acetylgalactal
PubChem CID 640125
IUPAC Name [(2R,3R,4R)-3,4-diacetyloxy-3,4-dihydro-2H-pyran-2-yl]methyl acetate
SMILES CC(=O)OCC1OC=CC(OC(C)=O)C1OC(C)=O

Specifications

Melting Point 34°C to 38°C
Color Yellow
Flash Point >110°C (235°F)
Quantity 1 g
Solubility Information Soluble in chloroform at 10mg/ml
Formula Weight 272.25
Physical Form Oil or Solid
Chemical Name or Material 3,4,6-Tri-O-acetyl-D-galactal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.