missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxytoluene, 98%

Product Code. 11462218
Change view
Click to view available options
Quantity:
5 g
25 g
50 g
100 g
Unit Size:
100g
25g
50g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462218 25 g 25g
11452218 5 g 5g
11359539 50 g 50g
11472218 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11462218 Supplier Thermo Scientific Alfa Aesar Supplier No. A15201.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6443-69-2 | C10H14O3 | 182.219 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6443-69-2
Molecular Formula C10H14O3
Molecular Weight (g/mol) 182.219
MDL Number MFCD00008397
InChI Key KCIZTNZGSBSSRM-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxytoluene, benzene, 1,2,3-trimethoxy-5-methyl, 5-methylpyrogallol trimethyl ether, 1,2,3-trimethoxy-5-methyl-benzene, toluene, 3,4,5-trimethoxy, 1,2,3-trimethoxy-5-methylbenzol, pubchem13665, acmc-209nm4, 3,4,5-trimethoxy-toluene, 3,4,5-trimethoxy toluene
PubChem CID 80922
ChEBI CHEBI:81354
IUPAC Name 1,2,3-trimethoxy-5-methylbenzene
SMILES CC1=CC(=C(C(=C1)OC)OC)OC

Specifications

Melting Point 34°C to 38°C
Density 1.082
Boiling Point 115°C to 116°C (4 mmHg)
Flash Point 74°C (165°F)
Refractive Index 1.523
Quantity 25 g
Beilstein 1953791
Formula Weight 182.22
Percent Purity 98%
Chemical Name or Material 3,4,5-Trimethoxytoluene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.