missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxyphenylacetic Acid, TRC

Product Code. 30450347
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30450347 50 g 50g
30449687 1 g 1g
30450022 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30450347 Supplier Toronto Research Chemicals Supplier No. TRCT79547550G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 951-82-6
Molecular Formula C11H14O5
Molecular Weight (g/mol) 226.23
Synonym (3,4,5-Trimethoxyphenyl)-acetic Acid, 3,4,5-Trimethoxybenzeneacetic Acid, 3,4,5-Trimethoxyphenylacetic Acid, NSC 130961, NSC 91017
IUPAC Name 2-(3,4,5-trimethoxyphenyl)acetic acid
SMILES COc1cc(CC(=O)O)cc(OC)c1OC

Specifications

Quantity 50 g
InChI Formula InChI=1S/C11H14O5/c1-14-8-4-7(6-10(12)13)5-9(15-2)11(8)16-3/h4-5H,6H2,1-3H3,(H,12,13)
Formula Weight 226.0841
Chemical Name or Material 3,4,5-Trimethoxyphenylacetic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.