missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzylamine, 96%

Product Code. 10038831 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10038831

Brand: Thermo Scientific Acros 351170250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18638-99-8 | C10H15NO3 | 197.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18638-99-8
Molecular Formula C10H15NO3
Molecular Weight (g/mol) 197.24
MDL Number MFCD00017150
InChI Key YUPUSBMJCFBHAP-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxybenzylamine, 3,4,5-trimethoxyphenyl methanamine, 1-3,4,5-trimethoxyphenyl methanamine, benzenemethanamine, 3,4,5-trimethoxy, 3,4,5-trimethoxyphenyl methylamine, 1-3,4,5-trimethoxyphenyl methanamine hydrochloride, pubchem16674, acmc-1c5ez, ksc495e6b, 3,4,5-trimethoxy benzylamine
PubChem CID 87736
IUPAC Name (3,4,5-trimethoxyphenyl)methanamine
SMILES COC1=CC(=CC(=C1OC)OC)CN

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1550g/mL
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)3C6H2CH2NH2
Refractive Index 1.5455 to 1.5485
Quantity 25 g
Specific Gravity 1.155
Formula Weight 197.24
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3, 4, 5-Trimethoxybenzylamine, 96%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4,5-Trimethoxybenzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.