missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzoic acid, 99%

Product Code. 10205290 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10205290 100 g Plastic bottle 100g
10706161 5 g Glass Bottle 5g
10336690 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10205290 Supplier Thermo Scientific Acros Supplier No. 139931000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118-41-2 | C10H12O5 | 212.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-41-2
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.2
MDL Number MFCD00002501
InChI Key SJSOFNCYXJUNBT-UHFFFAOYSA-N
Synonym eudesmic acid, gallic acid trimethyl ether, tri-o-methylgallic acid, trimethylgallic acid, benzoic acid, 3,4,5-trimethoxy, veratric acid, 5-methoxy, unii-v5c9h0sc9f, 3,4,5-trimethoxy-benzoic acid, 5-methoxy-veratric acid, v5c9h0sc9f
PubChem CID 8357
ChEBI CHEBI:454991
IUPAC Name 3,4,5-trimethoxybenzoic acid
SMILES COC1=CC(=CC(OC)=C1OC)C(O)=O

Specifications

CAS Min % 98.5
CAS Max % 100
Melting Point 168.0°C to 172.0°C
Color Colorless to Yellow
Boiling Point 225.0°C to 227.0°C (10.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula (CH3O)3C6H2CO2H
Quantity 100 g
Beilstein 10, 481
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ethanol,ethylether,chloroform
Formula Weight 212.2
Percent Purity 0.99
Physical Form Liquid
Chemical Name or Material 3, 4, 5-Trimethoxybenzoic acid, 0.99
Show More Show Less
compliance-icons
Product Identifier
  • 3,4,5-Trimethoxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.