missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzoic acid, 99+%

Product Code. 11472427
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11472427 500 g 500g
11462427 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11472427 Supplier Thermo Scientific Alfa Aesar Supplier No. A14274.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118-41-2 | C10H12O5 | 212.2 g/mol

3,4,5-Trimethoxybenzoic acid is used as intermediates in medicine and organic synthesis. It acts as a metabolite of trimebutine. It is widely used in making dyes and inks, photographic developers and used as astringents in medically.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5-Trimethoxybenzoic acid is used as intermediates in medicine and organic synthesis. It acts as a metabolite of trimebutine. It is widely used in making dyes and inks, photographic developers and used as astringents in medically.

Solubility
Soluble in ethanol, ethyl ether and chloroform. Slightly soluble in water.

Notes
Store away from oxidizing agents. Store in a cool, dry conditions in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-41-2
Molecular Formula C10H12O5
Molecular Weight (g/mol) 212.2
MDL Number MFCD00002501
InChI Key SJSOFNCYXJUNBT-UHFFFAOYSA-N
Synonym eudesmic acid, gallic acid trimethyl ether, tri-o-methylgallic acid, trimethylgallic acid, benzoic acid, 3,4,5-trimethoxy, veratric acid, 5-methoxy, unii-v5c9h0sc9f, 3,4,5-trimethoxy-benzoic acid, 5-methoxy-veratric acid, v5c9h0sc9f
PubChem CID 8357
ChEBI CHEBI:454991
IUPAC Name 3,4,5-trimethoxybenzoic acid
SMILES COC1=CC(=CC(OC)=C1OC)C(O)=O

Specifications

Melting Point 168°C to 171°C
Boiling Point 225°C to 226°C (10 mmHg)
Quantity 500 g
Beilstein 884655
Solubility Information Soluble in ethanol,ethyl ether and chloroform. Slightly soluble in water.
Formula Weight 212.2
Percent Purity ≥99%
Chemical Name or Material 3,4,5-Trimethoxybenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.