missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxybenzaldehyde, 98%

Product Code. 11497287
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11497287 500 g 500g
11477287 25 g 25g
11487287 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497287 Supplier Thermo Scientific Alfa Aesar Supplier No. A15038.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 86-81-7 | C10H12O4 | 196.202 g/mol

3,4,5-Trimethoxybenzaldehyde is used in the synthesis of trimethoprim and psychedelic phenethylamines. Also it can also be used in the production of plastic additives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5-Trimethoxybenzaldehyde is used in the synthesis of trimethoprim and psychedelic phenethylamines. Also it can also be used in the production of plastic additives.

Solubility
Soluble in water (1.49 mg/L at 25°C), alcohol, toluene, acetone, and methanol.

Notes
Air Sensitive, store away from air. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-81-7
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00003364
InChI Key OPHQOIGEOHXOGX-UHFFFAOYSA-N
PubChem CID 6858
IUPAC Name 3,4,5-trimethoxybenzaldehyde
SMILES COC1=CC(=CC(=C1OC)OC)C=O

Specifications

Melting Point 73°C to 77°C
Boiling Point 163°C to 165°C (10 mmHg)
Quantity 500 g
Beilstein 395163
Sensitivity Air Sensitive
Synonym benzaldehyde, 3,4,5-trimethoxy, unii-wl86yd76n6, tmba, benzaldehyde,3,4,5-trimethoxy, 3,4,5-trimethoxy benzaldehyde, 3,4,5-trimethoxy-benzaldehyde, pubchem8266, acmc-209qbj, 3,5-trimethoxybenzaldehyde, 2.3.4-trihyroxybenzaldehyde
Solubility Information Soluble in water (1.49mg/L at 25°C),alcohol,toluene,acetone,and methanol.
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material 3,4,5-Trimethoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.