missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 3,4,5-Trichlorothiophene-2-carboxylic acid, 98+%
Click to view available options
Quantity:
1 g
5 g
Description
Applications
It is a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store at room temperature. Keep away from strong oxidizing agents. Ensure proper ventilation.
It is a pharmaceutical intermediate.
Solubility
Insoluble in water.
Notes
Store at room temperature. Keep away from strong oxidizing agents. Ensure proper ventilation.
Specifications
Melting Point | 226°C to 228°C |
Quantity | 1 g |
Beilstein | 155175 |
Solubility Information | Insoluble in water. |
IUPAC Name | trichlorothiophene-2-carboxylic acid |
Formula Weight | 231.49 |
Percent Purity | ≥98% |
Chemical Name or Material | 3,4,5-Trichlorothiophene-2-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'