Learn More
3,4,5-Trichloropyridine, 98%, Thermo Scientific Chemicals
£42.70 - £658.00
Chemical Identifiers
CAS | 33216-52-3 |
---|---|
Molecular Formula | C5H2Cl3N |
Molecular Weight (g/mol) | 182.428 |
MDL Number | MFCD00051685 |
InChI Key | KKWRVUBDCJQHBZ-UHFFFAOYSA-N |
Synonym | pyridine, 3,4,5-trichloro, 3,4,5-trichloro-pyridine, pubchem11021, acmc-209hzc, ksc497k5t, 3,4,5-trichloropyridine, 3,4,5-trichloro pyridine, 3,4,5-trichloropyridine 1g |
PubChem CID | 3786531 |
IUPAC Name | 3,4,5-trichloropyridine |
SMILES | C1=C(C(=C(C=N1)Cl)Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11434304
|
Thermo Scientific Alfa Aesar
A12587.03 |
1 g |
£42.70
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11444304
|
Thermo Scientific Alfa Aesar
A12587.06 |
5 g |
£143.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11454304
|
Thermo Scientific Alfa Aesar
A12587.14 |
25 g | |||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
33216-52-3 | |
182.428 | |
KKWRVUBDCJQHBZ-UHFFFAOYSA-N | |
3786531 | |
C1=C(C(=C(C=N1)Cl)Cl)Cl |
C5H2Cl3N | |
MFCD00051685 | |
pyridine, 3,4,5-trichloro, 3,4,5-trichloro-pyridine, pubchem11021, acmc-209hzc, ksc497k5t, 3,4,5-trichloropyridine, 3,4,5-trichloro pyridine, 3,4,5-trichloropyridine 1g | |
3,4,5-trichloropyridine |
Specifications
33216-52-3 | |
213°C to 215°C | |
MFCD00051685 | |
pyridine, 3,4,5-trichloro, 3,4,5-trichloro-pyridine, pubchem11021, acmc-209hzc, ksc497k5t, 3,4,5-trichloropyridine, 3,4,5-trichloro pyridine, 3,4,5-trichloropyridine 1g | |
C1=C(C(=C(C=N1)Cl)Cl)Cl | |
182.428 | |
182.44 | |
3,4,5-Trichloropyridine |
70°C to 74°C | |
C5H2Cl3N | |
120542 | |
KKWRVUBDCJQHBZ-UHFFFAOYSA-N | |
3,4,5-trichloropyridine | |
3786531 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only