missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Tribromo-1H-pyrazole, 97%

Product Code. 11419902
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419902 25 g 25g
11409902 5 g 5g
11429902 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419902 Supplier Thermo Scientific Alfa Aesar Supplier No. A12031.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17635-44-8 | C3HBr3N2 | 304.767 g/mol

2,4,5-Tribromoimidazole(2,4,5-TBI) on condensation with sugar precursors yields 2,4,5-TBI nucleosides. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,4,5-Tribromoimidazole(2,4,5-TBI) on condensation with sugar precursors yields 2,4,5-TBI nucleosides. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water. Soluble in Chloroform.

Notes
Store in cool place. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17635-44-8
Molecular Formula C3HBr3N2
Molecular Weight (g/mol) 304.767
MDL Number MFCD00040248
InChI Key TXQKCKQJBGFUBF-UHFFFAOYSA-N
Synonym 3,4,5-tribromopyrazole, 1h-pyrazole, 3,4,5-tribromo, pyrazole, 3,4,5-tribromo, acmc-1c9yo, 3,4,5-tribromo pyrazole, ksc497i1h, #, 3,4,5-tribromo-1h-pyrazole 5g
PubChem CID 627674
IUPAC Name 3,4,5-tribromo-1H-pyrazole
SMILES C1(=C(NN=C1Br)Br)Br

Specifications

Melting Point 184°C to 188°C
Odor Odorless
Quantity 25 g
Beilstein 113501
Solubility Information Insoluble in water. Soluble in Chloroform.
Formula Weight 304.78
Percent Purity 97%
Chemical Name or Material 3,4,5-Tribromo-1H-pyrazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.