missing translation for 'onlineSavingsMsg'
Learn More

3-(4,5-Dimethyl-2-thazolyl)-2,5-diphenyl-2H-tetrazolium bromide, ∽98%, MP Biomedicals™

Product Code. 11319089 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
21g
This item is not returnable. View return policy

Product Code. 11319089

Brand: MP Biomedicals 0210222710

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 298-93-1
Molecular Formula C18H16BrN5S
Molecular Weight (g/mol) 414.33
MDL Number MFCD00011964,MFCD00066662
InChI Key AZKSAVLVSZKNRD-UHFFFAOYSA-M
Synonym thiazolyl blue, thiazolyl blue tetrazolium bromide, mmt tetrazolium, methylthiazoletetrazolium, mtt, thiazolyl blue monotetrazolium, thiazole blue, 3-4,5-dimethylthiazol-2-yl-2,5-diphenyltetrazolium bromide, mtt van, unii-euy85h477i
PubChem CID 64965
ChEBI CHEBI:53233
SMILES [Br-].CC1=C(C)N=C(S1)[N+]1=NC(=NN1C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color Orange-Yellow
Quantity 10 g
IUPAC Name 2-(3,5-diphenyltetrazol-2-ium-2-yl)-4,5-dimethyl-1,3-thiazole;bromide
Formula Weight 414.32
Percent Purity ∼98%
Physical Form Crystalline Powder
Chemical Name or Material 3-(4, 5-Dimethyl-2-thazolyl)-2, 5-diphenyl-2H-tetrazolium bromide
compliance-icons compliance-icons
Product Identifier
  • 3-(4,5-Dimethylthiazolyl-2)-2,5-diphenyl Tetrazolium Bromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 3-(4,5-Dimethylthiazolyl-2)-2,5-diphenylTetrazolium
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.