missing translation for 'onlineSavingsMsg'
Learn More

3,4,5,6-Tetrachloropyridine-2-carboxylic acid, 98%

Product Code. 11388756
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11388756 25 g 25g
11378756 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388756 Supplier Thermo Scientific Alfa Aesar Supplier No. L19450.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 10469-09-7 | C6HCl4NO2 | 260.88 g/mol

3,4,5,6-Tetrachloropyridine-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceutical and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,4,5,6-Tetrachloropyridine-2-carboxylic acid is an important raw material and intermediate used in organic synthesis, pharmaceutical and dyestuff.

Solubility
Soluble in DMSO, Methanol. Insoluble in water.

Notes
Incompatible with oxidizing agents. Store in a cool, dry conditions in well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10469-09-7
Molecular Formula C6HCl4NO2
Molecular Weight (g/mol) 260.88
MDL Number MFCD00185833
InChI Key GXFRQLQUKBSYQL-UHFFFAOYSA-N
Synonym 3,4,5,6-tetrachloropicolinic acid, tetrachloropicolinic acid, tetrachloropyridine-2-carboxylic acid, 2-pyridinecarboxylic acid, 3,4,5,6-tetrachloro, unii-7e9zn8ab09, 3,4,5,6-tetrachloro-2-pyridinecarboxylic acid, 2-pyridinecarboxylicacid, 3,4,5,6-tetrachloro, tetrachloropyridine-2-carboxylicacid, dsstox_cid_24509, dsstox_rid_80279
PubChem CID 82646
IUPAC Name 3,4,5,6-tetrachloropyridine-2-carboxylic acid
SMILES OC(=O)C1=NC(Cl)=C(Cl)C(Cl)=C1Cl

Specifications

Melting Point 171°C to 173°C
Quantity 25 g
Beilstein 479080
Solubility Information Soluble in DMSO,Methanol. Insoluble in water.
Formula Weight 260.89
Percent Purity 98%
Chemical Name or Material 3,4,5,6-Tetrachloropyridine-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.