missing translation for 'onlineSavingsMsg'
Learn More

3-(4-(2,3-Epoxypropoxy)phenyl)propionic Acid Methyl Ester, TRC

Product Code. 30384764
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30384764 500 mg
30385462 100 mg
30376685 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30384764 Supplier Toronto Research Chemicals Supplier No. TRCE811475500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81147-94-6
Molecular Formula C13H16O4
Molecular Weight (g/mol) 236.26
Synonym Esmolol Impurity 1
SMILES COC(=O)CCc1ccc(OCC2CO2)cc1

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C13H16O4/c1-15-13(14)7-4-10-2-5-11(6-3-10)16-8-12-9-17-12/h2-3,5-6,12H,4,7-9H2,1H3
Formula Weight 236.1049
Chemical Name or Material 3-(4-(2,3-Epoxypropoxy)phenyl)propionic Acid Methyl Ester
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.