missing translation for 'onlineSavingsMsg'
Learn More

3,4,2',3',4',5'-Hexachlorobiphenyl, TRC

Product Code. 30388917
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30388917 10 mg
30400749 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30388917 Supplier Toronto Research Chemicals Supplier No. TRCH29095810MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38380-08-4
Molecular Formula C12 H4 Cl6
Molecular Weight (g/mol) 360.88
Synonym 1,1'-Biphenyl, 2,3,3',4,4',5-hexachloro-, 2,3,3',4,4',5-Hexachloro-1,1'-biphenyl, 2,3,3',4,4',5-Hexachlorobiphenyl, 2,3,3',4,4',5-HxCB, 2,3,4,4',5,5'-Hexachlorobiphenyl, 2,3,4,5,3',4'-Hexachlorobiphenyl, 3,4,2',3',4',5'-Hexachlorobiphenyl, CB 156, PCB 156, 2,3,3',4,4',5-HexaCB
IUPAC Name 1,2,3,4-tetrachloro-5-(3,4-dichlorophenyl)benzene
SMILES Clc1ccc(cc1Cl)c2cc(Cl)c(Cl)c(Cl)c2Cl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H4Cl6/c13-7-2-1-5(3-8(7)14)6-4-9(15)11(17)12(18)10(6)16/h1-4H
Formula Weight 357.8444
Chemical Name or Material PCB No. 156
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.