missing translation for 'onlineSavingsMsg'
Learn More

3-[3-(Trifluoromethyl)benzoylamino]benzene boronic acid pinacol ester, 95%, Thermo Scientific™

Product Code. 15415145
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15415145 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15415145 Supplier Thermo Scientific Alfa Aesar Supplier No. H32372.ME
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1225069-85-1
Molecular Formula C20H21BF3NO3
MDL Number MFCD11844801
Quantity 500 mg
Synonym n-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-3-trifluoromethyl benzamide, 3-3-trifluoromethyl benzoylaminobenzene-boronic acid pinacol ester, 3-3-trifluoromethyl benzoylaminobenzene boronic acid pinacol ester, 3-3-trifluoromethyl benzoylamino benzene boronic acid pinacol ester, n-3-tetramethyl-1,3,2-dioxaborolan-2-yl phenyl-3-trifluoromethyl benzamide
InChI Key IUZSQEGKYZLEDM-UHFFFAOYSA-N
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC(=CC=C2)NC(=O)C3=CC(=CC=C3)C(F)(F)F
IUPAC Name N-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]-3-(trifluoromethyl)benzamide
Molecular Weight (g/mol) 391.197
PubChem CID 45588193
Formula Weight 391.2
Percent Purity 95%
Chemical Name or Material 3-[3-(Trifluoromethyl)benzoylamino]benzene boronic acid pinacol ester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.