missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Thiodipropionic acid, 99%

Product Code. 10295040 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
2.5kg
25g
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10295040 500 g Plastic bottle 500g
10205000 5 g Glass Bottle 5g
10183710 25 g Plastic bottle 25g
10391334 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10295040 Supplier Thermo Scientific Acros Supplier No. 138755000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 111-17-1 | C6H10O4S | 178.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-17-1
Molecular Formula C6H10O4S
Molecular Weight (g/mol) 178.2
MDL Number MFCD00002781
InChI Key ODJQKYXPKWQWNK-UHFFFAOYSA-N
Synonym 3,3'-thiodipropionic acid, thiodipropionic acid, propanoic acid, 3,3'-thiobis, bis 2-carboxyethyl sulfide, tyox a, thiodihydracrylic acid, 4-thiaheptanedioic acid, tdpa, sulfide, bis 2-carboxyethyl, 3,3'-thiodipropanoic acid
PubChem CID 8096
IUPAC Name 3-(2-carboxyethylsulfanyl)propanoic acid
SMILES C(CSCCC(=O)O)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 130.0°C to 133.0°C
Color White
Flash Point 128°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Plastic bottle
Linear Formula S(CH2CH2CO2H)2
Quantity 500 g
Beilstein 03, IV, 735
Merck Index 15, 9485
Solubility Information Solubility in water: 40g/L (25°C). Other solubilities: freely soluble in hot water, freely soluble in alcohol and acetone
Formula Weight 178.2
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3,3′-Thiodipropionic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3,3``-Thiodipropionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.