missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Thiodipropionic acid, 98%

Product Code. 11423447
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11423447 250 g 250g
11369236 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11423447 Supplier Thermo Scientific Alfa Aesar Supplier No. A17220.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 111-17-1 | C6H10O4S | 178.202 g/mol

3,3'-Thiodipropionic acid is used as an antioxidant for fats and other foodstuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,3′-Thiodipropionic acid is used as an antioxidant for fats and other foodstuffs.

Solubility
Soluble in acetone, ethanol. Slightly soluble in water.

Notes
Stable under ordinary conditions. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-17-1
Molecular Formula C6H10O4S
Molecular Weight (g/mol) 178.202
MDL Number MFCD00002781
InChI Key ODJQKYXPKWQWNK-UHFFFAOYSA-N
Synonym 3,3'-thiodipropionic acid, thiodipropionic acid, propanoic acid, 3,3'-thiobis, bis 2-carboxyethyl sulfide, tyox a, thiodihydracrylic acid, 4-thiaheptanedioic acid, tdpa, sulfide, bis 2-carboxyethyl, 3,3'-thiodipropanoic acid
PubChem CID 8096
IUPAC Name 3-(2-carboxyethylsulfanyl)propanoic acid
SMILES C(CSCCC(=O)O)C(=O)O

Specifications

Melting Point 127°C to 134°C
Linear Formula S(CH2CH2CO2H)2
Quantity 250 g
Beilstein 1210299
Merck Index 14,9332
Solubility Information Soluble in acetone,ethanol. Slightly soluble in water.
Formula Weight 178.21
Percent Purity 98%
Chemical Name or Material 3,3'-Thiodipropionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.