missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(3-Pyridyl)-L-alanine, 98%
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Description
3-(3-Pyridyl)-L-alanine is a antimycobacterial cyclodepsipeptide. PyrA is used to activate other aromatic acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-(3-Pyridyl)-L-alanine is a antimycobacterial cyclodepsipeptide. PyrA is used to activate other aromatic acids.
Solubility
Soluble in clear solution (0.3 gram in 2 ml DMF).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
3-(3-Pyridyl)-L-alanine is a antimycobacterial cyclodepsipeptide. PyrA is used to activate other aromatic acids.
Solubility
Soluble in clear solution (0.3 gram in 2 ml DMF).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Specifications
| Melting Point | 248°C to 252°C (decomposition) |
| Color | White |
| Beilstein | 4351583 |
| Solubility Information | Soluble in clear solution (0.3 gram in 2 ml DMF). |
| Quantity | 250 mg |
| Formula Weight | 166.18 |
| Percent Purity | 98% |
| Physical Form | Powder |
| Chemical Name or Material | 3-(3-Pyridyl)-L-alanine |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction