missing translation for 'onlineSavingsMsg'
Learn More

3,3'-[Oxybis(3,1-propanediyloxy)]bis[1-propanol], TRC

Product Code. 30424678
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30424678 10 mg 10mg
30411919 50 mg -
30419452 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30424678 Supplier Toronto Research Chemicals Supplier No. TRCO87029010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30242-05-8
Molecular Formula C12H26O5
Molecular Weight (g/mol) 250.33
Synonym 3,3'-[Oxybis(trimethyleneoxy)]di-1-propanol, NSC 361615
IUPAC Name 3,3'-((oxybis(propane-3,1-diyl))bis(oxy))bis(propan-1-ol)
SMILES OCCCOCCCOCCCOCCCO

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C12H26O5/c13-5-1-7-15-9-3-11-17-12-4-10-16-8-2-6-14/h13-14H,1-12H2
Formula Weight 250.18
Chemical Name or Material 3,3'-[Oxybis(3,1-Propanediyloxy)]Bis[1-Propanol]
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.