missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Methyl-1,2,4-oxadiazol-5-yl)piperidine, 98%, Thermo Scientific™

Product Code. 11439580
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439580 250 mg 250mg
11449580 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439580 Supplier Thermo Scientific Alfa Aesar Supplier No. H50883.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21767-12-4
Molecular Formula C11H15NO2
Molecular Weight (g/mol) 193.246
MDL Number MFCD07801211
InChI Key RLANOFAQNVDAQD-UHFFFAOYSA-N
Synonym 3-2-methoxyphenoxy pyrrolidine, 3-2-methoxy-phenoxy-pyrrolidine, s-3-2-methoxyphenoxy pyrrolidine, acmc-20apfb, 3-o-methoxyphenoxypyrrolidine, 3-2-methoxyphenoxy-pyrrolidine, pyrrolidine,3-2-methoxyphenoxy-, 3s
PubChem CID 14497355
IUPAC Name 3-(2-methoxyphenoxy)pyrrolidine
SMILES COC1=CC=CC=C1OC2CCNC2

Specifications

Quantity 250 mg
Formula Weight 167.21
Percent Purity 98%
Chemical Name or Material 3-(3-Methyl-1,2,4-oxadiazol-5-yl)piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.