missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Methoxyphenyl)propionic acid, 99%, Thermo Scientific™

Product Code. 15442212 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15442212 25 g 25g
15432212 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15442212 Supplier Thermo Scientific Acros Supplier No. 458110250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10516-71-9
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.20
MDL Number MFCD00014027
InChI Key BJJQJLOZWBZEGA-UHFFFAOYSA-N
Synonym 3-3-methoxyphenyl propionic acid, 3-3-methoxyphenyl propanoic acid, 3-m-methoxyphenyl propionic acid, 3-methoxybenzenepropanoic acid, benzenepropanoic acid, 3-methoxy, 3-3-methoxy-phenyl-propionic acid, 3-methoxy-benzenepropanoic acid, 3-3-methoxyphenyl propanoate, m-methoxyhydrocinnamate, 3-methoxydihydrocinnamate
PubChem CID 66336
IUPAC Name 3-(3-methoxyphenyl)propanoic acid
SMILES COC1=CC=CC(CCC(O)=O)=C1

Specifications

Melting Point 43.0°C to 46.5°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Quantity 25 g
Formula Weight 180.21
Percent Purity 99%
Chemical Name or Material 3-(3-Methoxyphenyl)propionic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.