missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Hydroxyphenyl)propionic Acid, TRC

Product Code. 30393221
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30393221 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393221 Supplier Toronto Research Chemicals Supplier No. TRCH94009010G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(3-Hydroxyphenyl)propionic Acid
CAS 621-54-5
Molecular Formula C9 H10 O3
Quantity 10 g
Synonym Benzenepropanoic acid, 3-hydroxy-, Hydrocinnamic acid, m-hydroxy- (7CI,8CI), 3-Hydroxybenzenepropanoic acid, 3-(3-Hydroxyphenyl)propanoic acid, 3-(3-Hydroxyphenyl)propionic acid, 3-(3'-Hydroxyphenyl)propionic acid, 3-(m-Hydroxyphenyl)propionic acid, 3-Hydroxydihydrocinnamic acid, 3-Hydroxyphenylpropionic acid, Dihydro-m-coumaric acid, MFCD 00002598, NSC 33135, NSC 39468, m-Hydroxybenzene propanoic acid, m-Hydroxyphenylpropionic acid, β-(3-Hydroxyphenyl)propionic acid, β-(m-Hydroxyphenyl)propionic acid
InChI Formula InChI=1S/C9H10O3/c10-8-3-1-2-7(6-8)4-5-9(11)12/h1-3,6,10H,4-5H2,(H,11,12)
SMILES OC(=O)CCc1cccc(O)c1
IUPAC Name 3-(3-hydroxyphenyl)propanoic acid
Molecular Weight (g/mol) 166.17
Formula Weight 166.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.