missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Furyl)acrylic Acid, TRC

Product Code. 30406564
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30406564 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30406564

Brand: Toronto Research Chemicals TRCF8652401G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(3-Furyl)acrylic Acid
CAS 39244-10-5
Molecular Formula C7H6O3
Quantity 1 g
Synonym 3-Furanacrylic Acid, 3-(3-Furanyl)-2-propenoic Acid, 3-(3-Furyl)acrylic Acid, 3-Furylacrylic Acid
InChI Formula InChI=1S/C7H6O3/c8-7(9)2-1-6-3-4-10-5-6/h1-5H,(H,8,9)
SMILES OC(=O)C=Cc1cocc1
IUPAC Name 3-(furan-3-yl)prop-2-enoic acid
Molecular Weight (g/mol) 138.12
Formula Weight 138.0317
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.