missing translation for 'onlineSavingsMsg'
Learn More

3-(3-Fluorobenzyl)-4-[(3-fluorobenzyl)oxy]benzaldehyde, TRC

Product Code. 30407853
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30407853 10 mg 10mg
30392935 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30407853 Supplier Toronto Research Chemicals Supplier No. TRCF59793510MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(3-Fluorobenzyl)-4-[(3-fluorobenzyl)oxy]benzaldehyde
CAS 1000370-24-0
Molecular Formula C21 H16 F2 O2
Quantity 10 mg
Synonym 4-[(3-Fluorophenyl)methoxy]-3-[(3-fluorophenyl)methyl]benzaldehyde
InChI Formula InChI=1S/C21H16F2O2/c22-19-5-1-3-15(11-19)9-18-10-16(13-24)7-8-21(18)25-14-17-4-2-6-20(23)12-17/h1-8,10-13H,9,14H2
SMILES Fc1cccc(COc2ccc(C=O)cc2Cc3cccc(F)c3)c1
IUPAC Name 4-[(3-fluorophenyl)methoxy]-3-[(3-fluorophenyl)methyl]benzaldehyde
Molecular Weight (g/mol) 338.347
Formula Weight 338.112
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.