missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dithiobis-L-Valine, TRC

Product Code. 30386126
Change view
Click to view available options
Quantity:
250 mg
1 g
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30386126 250 mg
30375398 1 g
30372964 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386126 Supplier Toronto Research Chemicals Supplier No. TRCD493795250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 113626-33-8
Molecular Formula C10H20N2O4S2
Molecular Weight (g/mol) 296.41
Synonym Penicillamine Disulphide Impurity
SMILES OC([C@@H](N)C(C)(C)SSC(C)([C@H](N)C(O)=O)C)=O

Specifications

InChI Formula InChI=1S/C10H20N2O4S2/c1-9(2,5(11)7(13)14)17-18-10(3,4)6(12)8(15)16/h5-6H,11-12H2,1-4H3,(H,13,14)(H,15,16)/t5-,6-/m1/s1
Quantity 250 mg
Formula Weight 296.0864
Shipping Condition Room Temperature
Chemical Name or Material 3,3'-Dithiobis-L-valine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.