missing translation for 'onlineSavingsMsg'
Learn More

3,3-Diphenylpropylamine 98%, ACROS Organics™

Product Code. 10774651 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10774651 100 g Glass bottle 100g
10286010 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10774651 Supplier Thermo Scientific Acros Supplier No. 159481000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 5586-73-2
Molecular Formula C15H17N
Molecular Weight (g/mol) 211.308
MDL Number MFCD00008202
InChI Key KISZTEOELCMZPY-UHFFFAOYSA-N
Synonym 3,3-diphenylpropylamine, benzenepropanamine, .gamma.-phenyl, 3-3-diphenylpropylamine, 3,3-diphenylpropan-1-amin, acmc-209lpx, 3.3-diphenylpropylamine, 3,3 diphenylpropylamine, 3,3-diphenyl propylamine, 3,3-diphenyl-propylamine, 3,3-diphenylpropylamine e
PubChem CID 79698
IUPAC Name 3,3-diphenylpropan-1-amine
SMILES C1=CC=C(C=C1)C(CCN)C2=CC=CC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (C6H5)2CHCH2CH2NH2
Refractive Index 1.5820 to 1.5840 (20°C, 589nm)
Quantity 100 g
Formula Weight 211.31
Percent Purity ≥97.5%
Physical Form Liquid After Melting
Chemical Name or Material 3, 3-Diphenylpropylamine, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.