missing translation for 'onlineSavingsMsg'
Learn More

3,3-Diphenylpropionic acid, 97%

Product Code. 15790789 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15790789 100 g Glass bottle 100g
10458450 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15790789 Supplier Thermo Scientific Acros Supplier No. 117421000
 Restricted Item
Estimated Shipment: 03-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 606-83-7 | C15H14O2 | 226.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 606-83-7
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.28
MDL Number MFCD00002717
InChI Key BZQGAPWJKAYCHR-UHFFFAOYSA-N
Synonym 3,3-diphenylpropionic acid, benzenepropanoic acid, .beta.-phenyl, 3,3-diphenyl-propionic acid, propionic acid, 3,3-diphenyl, .beta.-phenylbenzenepropanoic acid, beta,beta-diphenylpropionic acid, hydrocinnamic acid, .beta.-phenyl, beta-phenylbenzenepropanoic acid, hydrocinnamic acid, beta-phenyl, pubchem13618
PubChem CID 64798
IUPAC Name 3,3-diphenylpropanoic acid
SMILES OC(=O)CC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 150°C to 155°C
Color White to Yellow
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C6H5)2CHCH2CO2H
Quantity 100 g
Beilstein 09, 680
Solubility Information Solubility in water: slightly soluble
Formula Weight 226.27
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 3-Diphenylpropionic acid, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.