missing translation for 'onlineSavingsMsg'
Learn More

3,3-Diphenylpropionic acid, 97%

Product Code. 10458450 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10458450 25 g Glass bottle 25g
15790789 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10458450 Supplier Thermo Scientific Acros Supplier No. 117420250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 606-83-7 | C15H14O2 | 226.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 606-83-7
Molecular Formula C15H14O2
Molecular Weight (g/mol) 226.28
MDL Number MFCD00002717
InChI Key BZQGAPWJKAYCHR-UHFFFAOYSA-N
Synonym 3,3-diphenylpropionic acid, benzenepropanoic acid, .beta.-phenyl, 3,3-diphenyl-propionic acid, propionic acid, 3,3-diphenyl, .beta.-phenylbenzenepropanoic acid, beta,beta-diphenylpropionic acid, hydrocinnamic acid, .beta.-phenyl, beta-phenylbenzenepropanoic acid, hydrocinnamic acid, beta-phenyl, pubchem13618
PubChem CID 64798
IUPAC Name 3,3-diphenylpropanoic acid
SMILES OC(=O)CC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 150°C to 155°C
Color White to Yellow
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C6H5)2CHCH2CO2H
Quantity 25 g
Beilstein 09, 680
Solubility Information Solubility in water: slightly soluble
Formula Weight 226.27
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3, 3-Diphenylpropionic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 3,3-Diphenylpropionic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.