missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylpiperidine hydrochloride, 95%, Thermo Scientific™

Product Code. 10230100
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10230100 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10230100 Supplier Thermo Scientific Maybridge Supplier No. MAY00227DA
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 27832-58-2
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C7H16ClN
Quantity 1 g
Synonym 3,3-dimethylpiperidine hydrochloride, 3,3-dimethylpiperidine hcl, 3,3-dimethylpiperidine, chloride, piperidine, 3,3-dimethyl-, hydrochloride, 3,3-dimethylpiperidine-hydrogen chloride 1/1
InChI Key BXFKQBZPFQBCCO-UHFFFAOYSA-N
SMILES CC1(CCCNC1)C.Cl
IUPAC Name 3,3-dimethylpiperidine;hydrochloride
Molecular Weight (g/mol) 149.662
PubChem CID 18686627
Formula Weight 149.66
Percent Purity 95%
Chemical Name or Material 3, 3-Dimethylpiperidine hydrochloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.