missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylpiperidine, 95%, Thermo Scientific Chemicals

Catalog Number 11339969
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1193-12-0
C7H15N
113.20
MFCD00005995
CDODDZJCEADUQQ-UHFFFAOYSA-N
70942
3,3-dimethylpiperidine
CC1(C)CCCNC1
This item is not returnable. View return policy
1193-12-0
C7H15N
113.20
MFCD00005995
CDODDZJCEADUQQ-UHFFFAOYSA-N
70942
3,3-dimethylpiperidine
CC1(C)CCCNC1

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

 
TRUSTED_SUSTAINABILITY
Density 0.81
Boiling Point 134°C to 136°C
Flash Point 26°C (79°F)
Odor Amine-like
Refractive Index 1.4475
Quantity 250 mg
UN Number UN2733
Sensitivity Air Sensitive
Synonym piperidine, 3,3-dimethyl, 3,3-dimethyl-piperidine, 3,3-dimethyl piperidine, pubchem10316, acmc-1bo7w, 3,3-dimethylpiperdine
Formula Weight 113.2
Percent Purity 95%
Chemical Name or Material 3,3-Dimethylpiperidine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'