missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylglutaric Anhydride 99%, ACROS Organics™

Product Code. 10467680 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10467680 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10467680 Supplier Thermo Scientific Acros Supplier No. 116260050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4160-82-1
CAS Min % 98.5
CAS Max % 100.0
Melting Point 122°C to 126°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C7H10O3
MDL Number MFCD00006684
Quantity 5 g
Beilstein 17, 419
Synonym 3,3-dimethylglutaric anhydride, 2h-pyran-2,6 3h-dione, dihydro-4,4-dimethyl, 4,4-dimethyldihydro-2h-pyran-2,6 3h-dione, glutaric anhydride, 3,3-dimethyl, 3,3-dimethylglutaricanhydride, glutaric anhydride, beta,beta-dimethyl, acmc-1adve, dihydro-4,4-dimethyl-2h-pyran-2,6 3h-dione, 3,3-dimethyladipic anhydride, 3,3 dimethylglutaric anhydride
InChI Key HIJQFTSZBHDYKW-UHFFFAOYSA-N
SMILES CC1(CC(=O)OC(=O)C1)C
IUPAC Name 4,4-dimethyloxane-2,6-dione
Molecular Weight (g/mol) 142.154
PubChem CID 77813
Formula Weight 142.15
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material 3, 3-Dimethylglutaric anhydride, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.