missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylglutaric acid, 98%

Product Code. 10467490 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10467490 25 g Glass bottle 25g
10743651 100 g Glass bottle 100g
10321514 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10467490 Supplier Thermo Scientific Acros Supplier No. 116240250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4839-46-7 | C7H12O4 | 160.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4839-46-7
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
MDL Number MFCD00002716
InChI Key DUHQIGLHYXLKAE-UHFFFAOYSA-N
Synonym 3,3-dimethylglutaric acid, pentanedioic acid, 3,3-dimethyl, 3,3-dimethylglutarate, 3,3-dimethyl-pentanedioic acid, .beta.,.beta.-dimethylglutaric acid, 3,3-dimethyl-glutaric acid, 3,3-dimethylpentaneodioic acid, glutaric acid,3-dimethyl, pentanedioic acid,3-dimethyl, beta,beta-dimethylglutaric acid
PubChem CID 20984
ChEBI CHEBI:68502
IUPAC Name 3,3-dimethylpentanedioic acid
SMILES CC(C)(CC(O)=O)CC(O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 98.0°C to 102.0°C
Color White to Beige
Boiling Point 290.0°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HO2CCH2C(CH3)2CH2CO2H
Quantity 25 g
Beilstein 02, 684
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,ethanol and ether,sparingly soluble in benzene and chloroform,insoluble in low-boiling petroleum fractions
Formula Weight 160.17
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 3, 3-Dimethylglutaric acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.