missing translation for 'onlineSavingsMsg'
Learn More

3,3'-Dimethyldiphenylamine, 98%

Product Code. 11470387
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470387 5 g 5g
11460387 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470387 Supplier Thermo Scientific Alfa Aesar Supplier No. B21765.06
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 626-13-1 | C14H15N | 197.281 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-13-1
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.281
MDL Number MFCD00059315
InChI Key CWVPIIWMONJVGG-UHFFFAOYSA-N
Synonym di-m-tolylamine, 3,3'-dimethyldiphenylamine, m,m'-ditolylamine, dim-tolylamine, bis 3-methylphenyl amine, 3-methyl-n-3-methylphenyl aniline, benzenamine, 3-methyl-n-3-methylphenyl, di-m-tolyl-amine, 3,3'-ditolylamine, pubchem8963
PubChem CID 7016139
IUPAC Name 3-methyl-N-(3-methylphenyl)aniline
SMILES CC1=CC(=CC=C1)NC2=CC=CC(=C2)C

Specifications

Density 1.04
Boiling Point 200°C (24 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.6225
Quantity 5 g
Formula Weight 197.28
Percent Purity 98%
Chemical Name or Material 3,3'-Dimethyldiphenylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.