missing translation for 'onlineSavingsMsg'
Learn More

3-[3-(Dimethylamino)propoxy]benzonitrile, 97%, Thermo Scientific™

Product Code. 10604785
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10604785 5 g Amber Glass Bottle 5g
10357644 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10604785 Supplier Thermo Scientific Maybridge Supplier No. CC59416DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 209538-81-8
Molecular Formula C12H16N2O
Molecular Weight (g/mol) 204.273
MDL Number MFCD09064991
InChI Key GXMUQXKOVKMYIO-UHFFFAOYSA-N
Synonym 3-3-dimethylamino propoxy benzonitrile, 3-3-dimethylamino propoxy benzenecarbonitrile
PubChem CID 20991220
IUPAC Name 3-[3-(dimethylamino)propoxy]benzonitrile
SMILES CN(C)CCCOC1=CC=CC(=C1)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 204.27
Percent Purity 97%
Chemical Name or Material 3-[3-(Dimethylamino)propoxy]benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.