missing translation for 'onlineSavingsMsg'
Learn More

3-[3-(Dimethylamino)propoxy]benzaldehyde, 95%, Thermo Scientific™

£19.60 - £55.20

Chemical Identifiers

CAS 26815-13-4
Molecular Formula C12H17NO2
Molecular Weight (g/mol) 207.273
MDL Number MFCD09064988
InChI Key CMVPGTWBRBJOQR-UHFFFAOYSA-N
Synonym 3-3-dimethylamino propoxy benzaldehyde, benzaldehyde,3-3-dimethylamino propoxy, 3-3-dimethylamino-propoxy-benzaldehyde, 3-3-dimethylaminopropoxy benzaldehyde
PubChem CID 10727031
IUPAC Name 3-[3-(dimethylamino)propoxy]benzaldehyde
SMILES CN(C)CCCOC1=CC=CC(=C1)C=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10422834
View Documents Promotion Details
Thermo Scientific Maybridge
CC59404CB
250 mg Amber Glass Bottle
£19.60
250mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10652885
View Documents Promotion Details
Thermo Scientific Maybridge
CC59404DA
1 g Amber Glass Bottle
£55.20
1g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

26815-13-4
207.273
CMVPGTWBRBJOQR-UHFFFAOYSA-N
10727031
CN(C)CCCOC1=CC=CC(=C1)C=O
C12H17NO2
MFCD09064988
3-3-dimethylamino propoxy benzaldehyde, benzaldehyde,3-3-dimethylamino propoxy, 3-3-dimethylamino-propoxy-benzaldehyde, 3-3-dimethylaminopropoxy benzaldehyde
3-[3-(dimethylamino)propoxy]benzaldehyde

Specifications

26815-13-4
100.0
C12H17NO2
3-3-dimethylamino propoxy benzaldehyde, benzaldehyde,3-3-dimethylamino propoxy, 3-3-dimethylamino-propoxy-benzaldehyde, 3-3-dimethylaminopropoxy benzaldehyde
CN(C)CCCOC1=CC=CC(=C1)C=O
207.273
207.27
3-[3-(Dimethylamino)propoxy]benzaldehyde
94.5
95%
MFCD09064988
CMVPGTWBRBJOQR-UHFFFAOYSA-N
3-[3-(dimethylamino)propoxy]benzaldehyde
10727031
95%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications