missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethylacrylic acid, 98%

Product Code. 11432898
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11432898

Brand: Thermo Scientific Alfa Aesar A16865.36

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 541-47-9 | C5H8O2 | 100.117 g/mol

Chemical Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chemical Intermediates.

Solubility
Soluble in water, and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 541-47-9
Molecular Formula C5H8O2
Molecular Weight (g/mol) 100.117
MDL Number MFCD00004366
InChI Key YYPNJNDODFVZLE-UHFFFAOYSA-N
Synonym 3,3-dimethylacrylic acid, senecioic acid, 3-methylcrotonic acid, 3-methyl-2-butenoic acid, 2-butenoic acid, 3-methyl, senecic acid, beta,beta-dimethylacrylic acid, crotonic acid, 3-methyl, beta,beta-dimethacrylic acid, beta-methylcrotonic acid
PubChem CID 10931
ChEBI CHEBI:37127
IUPAC Name 3-methylbut-2-enoic acid
SMILES CC(=CC(=O)O)C

Specifications

Melting Point 66°C to 68°C
Density 0.969
Boiling Point 198°C to 200°C
Flash Point 93°C (199°F)
Quantity 500 g
UN Number UN3261
Beilstein 1720305
Solubility Information Soluble in water,and methanol.
Formula Weight 100.12
Percent Purity 98%
Chemical Name or Material 3,3-Dimethylacrylic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.