missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl Clobazam, TRC

Product Code. 30373000
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30373000 10 mg 10mg
30387052 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30373000 Supplier Toronto Research Chemicals Supplier No. TRCD46495510MG

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,3-Dimethyl Clobazam
Analyte or Component Names 3,3-Dimethyl Clobazam
Shipping Condition Room Temperature
Molecular Formula C18H17ClN2O2
Synonym Clobazam EP Impurity D
InChI Formula InChI=1S/C18H17ClN2O2/c1-18(2)16(22)20(3)14-10-9-12(19)11-15(14)21(17(18)23)13-7-5-4-6-8-13/h4-11H,1-3H3/i1D3,2D3,3D3
Solution Type Neat
SMILES O=C1N(C)C(C=CC(Cl)=C2)=C2N(C3=CC=CC=C3)C(C1(C)C)=O
Industry Type Pharmaceutical
Molecular Weight (g/mol) 328.79
Quantity 10 mg
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.