missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-3,3-Dimethyl-2-butylamine, 98%

Product Code. 11484533
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484533 5 g 5g
11474533 1 g 1g
11494533 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484533 Supplier Thermo Scientific Alfa Aesar Supplier No. A11530.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3850-30-4 | C6H16N | 102.20 g/mol

(±)-3,3-Dimethyl-2-butylamine is used to produce 1-pyridin-3-yl-3-(1,2,2-trimethyl-propyl)-thiourea. It is also used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(+/-)-3,3-Dimethyl-2-butylamine is used to produce 1-pyridin-3-yl-3-(1,2,2-trimethyl-propyl)-thiourea. It is also used as pharmaceutical intermediates.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Store away from acid chlorides and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3850-30-4
Molecular Formula C6H16N
Molecular Weight (g/mol) 102.20
MDL Number MFCD00008078
InChI Key DXSUORGKJZADET-YFKPBYRVSA-O
Synonym 1,2,2-trimethylpropylamine, 2-amino-3,3-dimethylbutane, 2-butanamine, 3,3-dimethyl, 3,3-dimethyl-2-aminobutane, 3,3-dimethyl-2-butanamine, 3-amino-2,2-dimethylbutane, propylamine, 1,2,2-trimethyl, 3,3-dimethyl-2-butylamine, 3,3-dimethylbut-2-ylamine, 3,3-dimethyl-2-butyl amine
PubChem CID 520907
IUPAC Name 3,3-dimethylbutan-2-amine
SMILES C[C@H]([NH3+])C(C)(C)C

Specifications

Density 0.755
Quantity 5 g
UN Number UN2733
Sensitivity Air Sensitive
Solubility Information Fully miscible in water.
Formula Weight 101.19
Percent Purity 98%
Chemical Name or Material (±)-3,3-Dimethyl-2-butylamine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.