missing translation for 'onlineSavingsMsg'
Learn More

3,3-Dimethyl-2-butanone, 96%, Thermo Scientific Chemicals

Product Code. 10477520 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass Bottle
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10477520 100 mL Glass Bottle 100mL
10103090 500 mL Glass Bottle 500mL
11974821 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10477520 Supplier Thermo Scientific Acros Supplier No. 131251000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-97-8
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00008846
InChI Key PJGSXYOJTGTZAV-UHFFFAOYSA-N
Synonym pinacolone, 3,3-dimethyl-2-butanone, tert-butyl methyl ketone, pinacolin, 2-butanone, 3,3-dimethyl, pinacoline, t-butyl methyl ketone, methyl tert-butyl ketone, pinakolin, 2,2-dimethylbutanone
PubChem CID 6416
IUPAC Name 3,3-dimethylbutan-2-one
SMILES CC(=O)C(C)(C)C

Specifications

Melting Point -52.5°C
Color Colorless to Yellow
Density 0.8000g/mL
Boiling Point 106.0°C
Flash Point 5°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass Bottle
Linear Formula (CH3)3CCOCH3
Refractive Index 1.3950 to 1.3980
Quantity 100 mL
Beilstein 01, 694
Merck Index 15, 7551
Specific Gravity 0.8
Solubility Information Solubility in water: very soluble. Other solubilities: soluble in alcohol,ether and acetone
Formula Weight 100.16
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3, 3-Dimethyl-2-butanone, 96%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3,3-Dimethyl-2-butanone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.